missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemsplash Polypropylene Lab/Visitors Coat Navy with Velcro Closure

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Description
Visitors Polypropylene Disposable Lab Coat
- Material: 30gsm Polypropylene Fabric
- Features:
- Velcro Closure
- With Fold Over Collar
- Elasticated Cuffs
- Ideal for Non-Hazardous Environments
- Ideal for Food Industry
- PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Specifications
Specifications
| Product Type | Visitor Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | Navy |
| Closure Type | Hook and Loop |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Particulates |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction