Learn More
Masterflex™ Polypropylene Male Luer Adapters
Product Code. 15272560
Change view
Click to view available options
For Use With:
Fits Male Luer Lock to 1/16 in. I.D.
Fits Male Luer Lock to 1/8 in. I.D.
Fits Male Luer Lock to 3/32 in. I.D.
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15272560 Fits Male Luer Lock to 3/32 in. I.D. Pack of 25
15252400 Fits Male Luer Lock to 1/16 in. I.D. Pack of 25
15129222 Fits Male Luer Lock to 1/8 in. I.D. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15272560 Supplier Masterflex™ Supplier No. 4551824
  • ONLINE EXCLUSIVE PRICE
    20.76 €
    19.67 € / Pack of 25
    Save 1.09 €
    5% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
Complete your luer fittings systems with these premium connectors. Cole-Parmer™ Polypropylene Male Luer Adapters are manufactured in compliance with ISO 594-1 standards, free from contaminants and fully traceable. Ideal for a variety of critical applications, such as syringes or small tubing.
  • Manufactured of animal-free natural polypropylene
  • All parts meet tight dimensional tolerances for increased reliability and process purity
  • Choose from a range of sizes, with a variety of connection options
  • Good chemical resistance. Max temperature 87°C  (190°F)
  • Sterilize by ethylene oxide or gamma irradiation (not recommended for autoclaving)
  • All female ports may be used with either slip or luer lock connections
  • Plugs and caps are also available
Specifications
Material Polypropylene
For Use With Fits Male Luer Lock to 3/32 in. I.D.
For Use With (Application) Ideal for a variety of applications, including syringes or small tubing
Packaging Quantity 25 Pack
Product Type Luer Adapter