Learn More
Masterflex™ Polypropylene Male Luer Fitting
Product Code. 15353312
Change view
Click to view available options
For Use With:
Fits Male Luer Plug
Fits Male Luer x 1/18 in. hose barb
Fits Male luer with lock ring x female luer coupler
Fits Snap Luer Lock Rings
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15353312 Fits Male Luer x 1/18 in. hose barb Pack of 25
15252560 Fits Male Luer Plug Pack of 25
15330491 Fits Male luer with lock ring x female luer coupler Pack of 25
15313322 Fits Snap Luer Lock Rings Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15353312 Supplier Masterflex™ Supplier No. 4551844
Add to basket
Add to basket
Specifications
Material Polypropylene
For Use With Fits Male Luer x 1/18 in. hose barb
For Use With (Application) Ideal for a variety of applications, including syringes or small tubing
Packaging Quantity 25 Pack
Product Type Luer Fitting