Learn More
Masterflex™ Polypropylene Premium Luer Fitting
Product Code. 15296618 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
Fits 1/16 in. hose barb
Fits 3/16 in. hose barb
Fits 3/32 in. hose barb
Unit Size:
Each
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
15296618 Fits 3/16 in. hose barb Each
14889980 Fits 1/16 in. hose barb Pack of 25
13546720 Fits 3/32 in. hose barb Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15296618 Supplier Masterflex™ Supplier No. 4551808
  • ONLINE EXCLUSIVE PRICE
    21.15 €
    19.67 € / Pack of 1
    Save 1.48 €
    6% Off
View all promotions
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Complete your luer fittings systems with these premium connectors. Cole-Parmer™ Polypropylene Male Luer Fitting with Lock Rings are manufactured in compliance with ISO 594-1 standards, free from contaminants and fully traceable. Ideal for a variety of critical applications, such as syringes or small tubing.
  • Manufactured of animal-free natural polypropylene
  • All parts meet tight dimensional tolerances for increased reliability and process purity
  • Choose from a range of sizes, with a variety of connection options
  • Good chemical resistance. Max temperature 87°C  (190°F)
  • Sterilize by ethylene oxide or gamma irradiation (not recommended for autoclaving)
  • All female ports may be used with either slip or luer lock connections
  • Plugs and caps are also available
Specifications
Material Polypropylene
For Use With Fits 3/16 in. hose barb
For Use With (Application) Ideal for a variety of applications, including syringes or small tubing
Packaging Quantity 25 Pack
Product Type Luer Fitting