Learn More
Masterflex™ Polypropylene Male Pipe Adapter Barbed Fittings
Product Code. 15277536 Shop All Masterflex Products
Change view
Click to view available options
Thread Size:
1/4 in. MNPT
1/8 in. MNPT
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
15277536 1/8 in. MNPT Pack of 10
15202608 1/4 in. MNPT Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15277536 Supplier Masterflex™ Supplier No. 4062122
  • ONLINE EXCLUSIVE PRICE
    20.00 €
    18.73 € / Pack of 10
    Save 1.27 €
    6% Off
View all promotions
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With 1/8 x 5/32 in. Tubing I.D.
Packaging Quantity 10 Pack
Thread Size 1/8 in. MNPT
Product Type Male Pipe Adapter