missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Micro Tubes Cap Insert

Product Code. 10324851 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10324851 Yellow Pack of 500
10548931 Red Pack of 500
10028960 Green Pack of 500
10480243 Blue Pack of 500
15527851 White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10324851 Supplier BRAND™ Supplier No. 780724
  • ONLINE EXCLUSIVE PRICE
    29.32 €
    20.00 € / Pack of 500
    Save 9.32 €
    31% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polypropylene Micro Tubes Cap Insert for use with all non-tamper evident screw caps allows for easy identification.

  • For micro tubes with attached or bulk screw caps
  • Suitable for use with non-tamper evident caps
TRUSTED_SUSTAINABILITY

Specifications

Product Type Microtube Cap Insert
Material Polypropylene
Color Yellow
Temperature Range (Metric) -196°C to +121°C
For Use With (Equipment) Microtubes, Screw Caps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.