missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microcentrifuge Tubes without Cap

Product Code. 11538252 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
1.5 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11538252 1.5 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11538252
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Use these micro centrifuge tubes for a variety of applications. Fisherbrand™ Polypropylene Microcentrifuge Tubes without Cap are ideal for handling needs where a snap cap can interfere.

  • Conical
  • 1.5mL

Specifications

Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
For Use With (Equipment) Microcentrifuge
No. per Pack 1000/Pk.
Packaging 1000/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.