missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Makro II Natural Pipet Tip

Product Code. 11557462 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1000 to 5000 μL
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11557462 1000 to 5000 μL Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11557462
Only null left
Add to basket
Edge
Add to basket

Use these pipet tips with all commercially available pipets. Fisherbrand™ Polypropylene Polypropylene Makro II Natural Pipet Tips feature uniform high quality and precision, a perfect fit for manual or electronic, single or multichannel pipets

  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice

Specifications

Volume (Metric) 1000 to 5000 μL
Color Natural
Autoclavable Autoclavable
For Use With (Equipment) Sartorius Biohit, Labmate, Socorex
Product Line Makro II
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.