Learn More
Fisherbrand™ Polypropylene Pipet Tip G
Product Code. p-8012524 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 10 μL
Unit Size:
Pack of 1000
Pack of 192
Product Code. 11577442
Only null left
Add to basket
Edge
Add to basket
Make pipetting much simpler and safe for even the most sensitive applications. Fisherbrand™ Polypropylene Pipet Tips G are perfectly fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric) 0.1 to 10 μL
Color Crystal
Autoclavable Autoclavable
Material Polypropylene