Learn More
Witeg™ Polypropylene Pipet Racks
Product Code. 10591973
Change view
Click to view available options
Positions:
94
Unit Size:
Each
Product Code. Positions Unit Size
10591973 94 Each
1 options
Product Code. 10591973 Supplier Witeg™ Supplier No. 7 081 001
  • ONLINE EXCLUSIVE PRICE
    48.18 €
    40.02 € / Pack of 1
    Save 8.16 €
    16% Off
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specially designed to prevent any damage to the pipet tip. Witeg™ Polypropylene Pipet Racks hold up to 96 pipets.
  • Polypropylene
  • Residual liquids are retained by reinforced rim
Specifications
Holds Pipettes
Material Polypropylene
Positions 94
For Use With (Equipment) Pipets
Quantity 1 Pack
Product Type Pipet Rack