missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Protective Coveralls, 4515 Series

Product Code. 12902275 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
194 to 200 cm
‎186 to 194 cm
‎200 to 206 cm
Unit Size:
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Height (English) Height (Metric) Unit Size
12902275 Medium 66 to 69 in. 167 to 176 cm Pack of 20
12912275 Large 69 to 71 in. 174 to 181 cm Pack of 20
12932275 2X-Large 73 to 76 in. ‎186 to 194 cm Pack of 20
12992265 Small 64 to 67 in. 164 to 170 cm Pack of 20
12942275 3X-Large 76 to 78 in. 194 to 200 cm Pack of 20
12952275 4X-Large 78 to 81 in. ‎200 to 206 cm Pack of 20
12922275 X-Large 70 to 74 in. 179 to 187 cm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12902275 Supplier 3M™ Supplier No. 7000089612
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides PPE Directive Category III, Type 5/6 protection. The Protective Coverall 4515 Series protect from hazardous dusts such as asbestos fibers.

  • Breathable material reduces heat build-up and discomfort
  • Excellent barrier against dry particles and limited liquid splash (CE Type 5/6)
  • Robust, highly breathable 50gsm SMS material
  • Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
  • Two-way zipper with fully sealable storm flap
  • Two-panel hood design
  • Elasticated wrists, waist, and ankles for increased comfort and freedom of movement
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Medium
Gender Unisex
Material SMS Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class PPE Directive Category III, Type 5/6
Model 4515W
Size (English) Chest 36 to 39 in.
Size (Metric) Chest 92 to 100 cm
Height (English) 66 to 69 in.
Height (Metric) 167 to 176 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.