missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Polypropylene Reagent Bottles

Product Code. 15201179
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15201179 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15201179 Supplier Vitlab™ Supplier No. 101889
  • ONLINE EXCLUSIVE PRICE
    83.55 €
    71.09 € / Pack of 10
    Save 12.46 €
    14% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these bottles for long-term storage of liquids. VITLAB™ Polypropylene Reagent Bottles feature a wider mouth for simple filling of powders and paste-like materials.

  • Transparent Reagent Bottles
  • Good chemical resistance
  • With polypropylene screw cap
  • Autoclavable at 121°C (2 bar) according to DIN EN 285
  • 5000mL size features one handle for easy transport and pouring
  • Suitable for foodstuffs
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Transparent
For Use With (Application) Ideal for powders and paste-like materials
Includes Screw cap
Material Polypropylene
No. per Pack 10 Pack
Closure Included Yes
Mouth Wide
Height (Metric) 204 mm
Closure Material Polypropylene
Diameter (Metric) 53 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.