missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JG Finneran Associates™ Polypropylene Screw Thread Closures
Specifications
Specifications
| Color | Black |
| Material | Polypropylene |
| For Use With (Application) | For use with all autosampler |
| Quantity | 1000/Pk. |
| Closure Size | 8-425 |
| For Use With (Equipment) | 13 x 32 Screw Thread Vials with 8 to 425 mm Neck Finish |
| Product Type | Screw Cap |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?