Learn More
Chemsplash Polypropylene Socks (Pair) Elastic at Opening
GREENER_CHOICE
Product Code. 30058350
Change view
Click to view available options
Dimensions (L x W):
36 x 18 cm
Unit Size:
Pack of 250
Product Code. Dimensions (L x W) unitSize
30058350 36 x 18 cm Pack of 250
1 options
Product Code. 30058350 Supplier Chemsplash Supplier No. 2787.400.999
Add to basket
Add to basket
This product is suitable for use in BRC approved work areas and for the Pharmaceutical Industry.
40GSM Polypropylene Boot
  • 40GSM Polypropylene Material
  • Elastic at Opening
  • 36 x 18 cm
  • 10 pairs/pack
  • PFAS Free
Description: This product is designed for use in BRC approved work areas and the pharmaceutical industry. Made from 40GSM polypropylene material, it features an elasticated opening for a secure fit. The size is 36 x 18 cm with 10 pairs per pack. This boot is PFAS free and ensures safety and compliance with industry standards.
Specifications
Size Universal
Material Polypropylene
Latex Free Yes
Coating Polypropylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Plain
Dimensions (L x W) 36 x 18 cm
Weight (Metric) 40 gsm
Resistance Dirt
Product Type Shoe Covers