Learn More
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector
Product Code. 12393369
Change view
Click to view available options
Thread Size:
GL14
GL18
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
12393369 GL14 Pack of 10
12303379 GL18 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12393369 Supplier Lenz Laborglasinstrumente™ Supplier No. 01350000
  • ONLINE EXCLUSIVE PRICE
    16.02 €
    12.37 € / Pack of 10
    Save 3.65 €
    22% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For connecting screw caps with bore. Lenz Laborglasinstrumente™ Plastic Straight Hose Connectors are made from polypropylene.
  • Autoclavable
  • Available in two different thread sizes
  • Silicone sealing
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With For screw caps with bore
Packaging Quantity 10 Pack
Thread Size GL14
Shape Straight
Product Type Straight Hose Connection