missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Straight Tubing Adapters

Product Code. 11868321 Shop All Brand Products
Change view
Click to view available options
For Use With:
3 to 5 mm I.D. and 6 to 10 mm I.D.
3 to 5 mm I.D. and 9 to 13 mm I.D.
6 to 10 mm I.D. and 9 to 13 mm I.D.
Overall Length (Metric):
55 mm
59 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Overall Length (Metric) Unit Size
11868321 6 to 10 mm I.D. and 9 to 13 mm I.D. 59 mm Pack of 10
11858321 3 to 5 mm I.D. and 6 to 10 mm I.D. 55 mm Pack of 10
11878321 3 to 5 mm I.D. and 9 to 13 mm I.D. 55 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11868321 Supplier BRAND™ Supplier No. 152615
  • ONLINE EXCLUSIVE PRICE
    19.35 €
    17.50 € / Pack of 10
    Save 1.85 €
    9% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from polypropylene. BRAND™ Straight Tubing Adapters connect tubing of different diameters.

  • Manufactured from polypropylene
  • Connects tubing of different diameters
  • Translucent
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Overall Length (Metric) 59 mm
For Use With 6 to 10 mm I.D. and 9 to 13 mm I.D.
For Use With (Application) Connecting tubing of different inner diameters
Packaging Quantity 10 Pack
Shape Straight
Color Translucent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.