missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Translucent Polypropylene Straight Tubing Connectors

Product Code. 11833970 Shop All Brand Products
Change view
Click to view available options
Overall Length (Metric):
47 mm
55 mm
57 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Unit Size
11833970 55 mm Pack of 10
11894140 47 mm Pack of 10
10737391 57 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11833970 Supplier BRAND™ Supplier No. 152710
  • ONLINE EXCLUSIVE PRICE
    17.60 €
    15.24 € / Pack of 10
    Save 2.36 €
    13% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from translucent polypropylene. BRAND™ Translucent Polypropylene Straight Tubing Connectors are ridged to fit a variety of tubing diameters.

  • Manufactured from translucent polypropylene
  • Ridged to fit a variety of tubing diameters
  • Straight shape
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Polypropylene
Overall Length (Metric) 55 mm
For Use With 6 to 10mm tubing I.D.
Packaging Quantity 10 Pack
Shape Straight
Color Translucent
Product Type Straight Tubing Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.