Learn More
Kartell™ Polypropylene T-Shaped Untapered Connectors
BIOPHARMA
Obtain a stable connection and prevent slipping. Kartell™ Polypropylene ‘T’ Shaped Untapered Connectors are excellent for chemical and temperature resistance. The Connectors have serrations to hold hoses tightly and prevent slipping.
Brand:  Kartell™ 7052000
Product Code. 12000592
  • ONLINE EXCLUSIVE PRICE
    12.65 €
    8.80 € / Pack of 10
    Save 3.85 €
    30% Off
Only null left
In Stock
Add to basket
Description
Description
  • Chemical and temperature resistance
  • Serrations to hold hoses tightly
  • Polypropylene