Learn More
Kartell™ Polypropylene Tall-Form Measuring Cylinders
Manufactured from highly translucent polypropylene for durability, clarity and excellent chemical resistance. Autoclavable Kartell™ Polypropylene Tall Form Cylinders with Molded Graduations provide an economical and safe solution for measuring and mixing liquids in the laboratory.
Brand:  Kartell™ 0107500
Product Code. 11368513
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Manufactured from highly translucent polypropylene for durability, clarity and excellent chemical resistance. Autoclavable Kartell™ Polypropylene Tall Form Cylinders with Molded Graduations provide an economical and safe solution for measuring and mixing liquids in the laboratory.
  • Permanent molded graduations provide clarity and reproducibility
  • Autoclavable at 121°C for 20 minutes
  • Withstands 100°C of continuous work
  • Pentagon base for stability
  • Class B, Tall form
  • Complies with ISO 6706-1981(E) and BS 5404 Part 2 1977
  • Suitable for foodstuff