missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Thread Adapter

Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.

Brand:  Buerkle™ 0600-3004

Additional Details : Weight : 0.03515kg

 View more versions of this product

Product Code. 10014990

  • 18.45 € / Each

Estimated Shipment: 06-12-2023
to see stock.
Add to basket

Description

Description

  • Constructed of polypropylene
  • Interchange the connections to different adapters
Specifications

Specifications

Thread Adapter
Pump-it Container Pump
Green
Polypropylene
External: 2 in. coarse Mauser - 64 mm (BSI)
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers