missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, MP Biomedicals™

Product Code. 11431981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 L
Unit Size:
100mL
4 L
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431981 4 L 4 L
11416601 50 mL 50mL
11426601 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431981 Supplier MP Biomedicals Supplier No. 0219472404
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylenesorbitan monolaurate, Tween 20
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Oily Liquid
Density 1.095 g/mL
Quantity 4 L
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Chemical Name or Material Polysorbate 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.