missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, MP Biomedicals™

Product Code. 11470645 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470645 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470645 Supplier MP Biomedicals Supplier No. 0210316891
  • ONLINE EXCLUSIVE PRICE
    112.00 €
    106.05 € / 1 L
    Save 5.95 €
    5% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Derived from sorbitan ester. Polysorbate 20, MP Biomedicals is a nonionic surfactant that is soluble/dispersable in water. It is typically used for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications.

Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been used as a blocking agent on nitrocellulose.

Specifications

CAS 9005-64-5
Physical Form Liquid
Boiling Point >100°C (literature)
Density 1.095g/mL (Lit.)
Concentration Working Concentration: As a blocking agent on nitrocellulose at a typical concentration of 0.05%. Used at concentration of 2% for extraction of membrane-bound proteins.
pH 5 to 7 (1% aq. soln.) (literature)
Quantity 1 L
Refractive Index n20/D 1.468 (Lit.)
Specific Gravity 1.08-1.13
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Viscosity 400 cps at 25°C (Lit.)
MDL Number MFCD00165986
Synonym Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate, Glycosperse L-20X, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, TWEEN™, 20, PEG(20)sorbitan monolaurate
Solubility Information Soluble/miscible in water (1g/20 mL) to give a clear yellow solution; miscible with alcohol, dioxane, and ethyl acetate; practically insoluble in liquid paraffin and fixed oils (such as mineral oil).
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Molecular Weight (g/mol) 522.68
PubChem CID 443314
Formula Weight 1228
Concentration or Composition (by Analyte or Components) 0.06mM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.