missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, MP Biomedicals™

Product Code. 11421981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421981 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421981 Supplier MP Biomedicals Supplier No. 02194724.5
  • ONLINE EXCLUSIVE PRICE
    78.60 €
    74.55 € / 500mL
    Save 4.05 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions.

Tweens™ are a series of nonionic surfactants derived from sorbitan esters.

  • They are soluble or dispersible in water but differ widely in organic and oil solubilities.
  • Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.
  • Polysorbate 20 is nonionic surfactants derived from sorbitan ester.

Specifications

CAS 9005-64-5
Density 1.095g/mL
Quantity 500 mL
Physical Form Clear, Yellow to Amber Viscous Liquid
Refractive Index n20/D 1.468(lit.)
Specific Gravity 1.08–1.13
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MDL Number MFCD00165986
Viscosity 400 cps
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
Solubility Information Soluble/miscible in water to give a clear yellow solution; miscible with alcohol, dioxane, and ethyl acetate.
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Molecular Weight (g/mol) 522.68
PubChem CID 443314
Formula Weight about 1,228g/mol
Grade Cell Culture Reagent
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 0.06 mM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.