missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20 MP Biomedicals

Product Code. 11433601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433601 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433601 Supplier MP Biomedicals Supplier No. 0210316880
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 9005-64-5
Density 1.095g/mL
Quantity 100 mL
Assay Percent Range 55%
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MDL Number MFCD00165986
Synonym Polysorbate 20, Glycosperse L-20X, Polyoxyethylene (20) Sorbitan Monolaurate
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Molecular Weight (g/mol) 522.68
PubChem CID 443314
Chemical Name or Material Polysorbate 20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.