missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™

Product Code. 10381435 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Steel Drum
Unit Size:
1 L
19 L
20mL
4 L
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10381435 200 mL Narrow Mouth Amber Glass Bottle Each
15528264 20 mL Narrow Mouth Amber Glass Bottle 20mL
15548264 1 L Narrow Mouth Amber Glass Bottle 1 L
15538264 4 L Narrow Mouth Amber Glass Bottle 4 L
15558264 19 L Steel Drum 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10381435 Supplier Avantor J.T.Baker Supplier No. 411604
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 200 mL
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.11kg
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.