missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20

Product Code. 11388116
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388116 500 mL 500mL
11378116 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388116 Supplier Thermo Scientific Alfa Aesar Supplier No. L15029.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 20 is used as a wetting agent in flavored mouth drops. It acts as a washing agent in Western blots and ELISAs and prevents antibody binding. It is a solubilizing agent of membrane proteins. In pharmaceutical chemistry, it is used as an excipient to stabilize emulsions and suspensions. Further, it is used as a dispersing agent, stabilizer, lubricator and also used in creams, salves, ointments, protective creams, balms, pomades, lipsticks, mascaras and glosses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
PubChem CID 443314
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Boiling Point >100°C
Density 1.105
Quantity 500 mL
Flash Point >110°C (230°F)
Refractive Index 1.47
Solubility Information Miscible with water.
Chemical Name or Material Polysorbate 20

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.