missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 60 (Techincal Grade), TRC

Product Code. 30421576
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421576 Supplier Toronto Research Chemicals Supplier No. TRCP6894301G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polysorbate 60
CAS 9005-67-8
Molecular Formula C22H42O8
Quantity 1 g
Synonym Sorbitan Monostearate Polyoxyethylene Derivs., Admul T 60K, Ahco DFS 100, Ahco DFS 149, Armotan PMS 20, Atlas G 1036, Crill 8, Crill 9, Crill S 8, Crillet 3, Crillet 31, Disponil SMS 120F1, Drewpone 60, Durfax 60K, E 435, Emasol 3130, Emasol S 120V, Emerest 2654, Emsorb 6905, Emsorb 6906, Ethoxylated Sorbitan Monostearate, Ethsorbox S, Eumulgin SMS 20, Glycosperse S 20, Ionet T 60C, Kaopan TW-S 120, Kolliphor PS 60, Kotilen S 1, Lamesorb SMS 20, Lumisorb PSMS 20, MS 55F, Montanox 60, Montanox 60DF, Newcol 65, Nikkol TS 10, Nikkol TS 106
InChI Formula InChI=1S/C22H42O8/c1-2-3-4-5-6-7-8-9-10-11-20(26)29-16-18(25)21-22(28-15-13-24)19(17-30-21)27-14-12-23/h18-19,21-25H,2-17H2,1H3/t18-,19+,21-,22-/m1/s1
SMILES OCCO[C@H]1[C@H](OCCO)CO[C@@H]1[C@@H](COCCOC(CCCCCCCCCCCCCCCCC)=O)OCCO
Molecular Weight (g/mol) 434.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.