missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Colorimetric 8 Cell Tray

Product Code. 10441551
Change view
Click to view available options
Diameter (Metric):
15 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10441551 15 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10441551 Supplier Kartell™ Supplier No. 0035700
  • ONLINE EXCLUSIVE PRICE
    20.00 €
    18.58 € / Pack of 10
    Save 1.42 €
    7% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kartell™ Colorimetric 8 Cell Tray is constructed of polystyrene for durability and is best suited for colorimetric assay.

  • CE Marked, IVD Certified
  • 95mm long and 57mm wide
  • For colorimetric assay

Specifications

Material Polystyrene
Shape Rectangular
Type Tray
Length (Metric) 95 mm
Width (Metric) 57 mm
Height (Metric) 2 mm
Diameter (Metric) 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.