missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene latex microsphere, 0.20 micron, 2.5 wt% dispersion in water

Product Code. 15496505
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496505 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Artikelnummer. 15496505 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. 042713.4Y
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Polystyrene latex microspheres are utilized in flow cytometry, fluorescence microscopy, and as calibration particles. It finds a use in the detection of trace amounts of antigens or antibodies in serum, urine, and cerebro-spinal fluids. It is also employed as a stimulus responsive particulate emulsifier for oil-in-water emulsion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spezifikation

Color White
CAS 9003-53-6
Physical Form Dispersion
Quantity 15 mL
Odor Odorless
Molecular Formula (C8H8)n
Particle Size 0.20 Micron
MDL Number MFCD00084450
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
Solubility Information Soluble in methylethylketone,benzene and toluene. Slightly soluble in diethyl ether,n-octanol and acetone. Insoluble in water.
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES *-CC(-*)C1=CC=CC=C1
IUPAC Name styrene
Molecular Weight (g/mol) 104.15
PubChem CID 7501
ChEBI CHEBI:27452
Concentration or Composition (by Analyte or Components) 2.5 wt% dispersion in water
Chemical Name or Material Polystyrene latex microsphere
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.