Learn More
Polystyrene latex microsphere, 0.75 micron, 2.5 wt% dispersion in water, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar 042742.4Y
Quantity 15mL
Product Code. 15476515
  • 476.00 € / 15mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
9003-53-6
MFCD00008612,MFCD00084450
Fully miscible with water.
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene
7501
Odorless
Polystyrene latex microsphere
C8H8
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
15mL
C=CC1=CC=CC=C1
104.15
CHEBI:27452
0.75 Micron
Product Suggestions
Product Suggestions
Videos
SDS Documents