missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene latex microsphere, 1 micron, 10 wt% dispersion in water

Product Code. 15446415
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446415 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446415 Supplier Thermo Scientific Alfa Aesar Supplier No. 041917.7C
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Use in protein binding; size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9003-53-6
Physical Form Dispersion
Quantity 15 mL
Odor Odorless
Molecular Formula (C8H8)n
Particle Size 1 Micron
MDL Number MFCD00084450
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
Solubility Information Soluble in Benzene,chloroform,carbon tetrachloride,cyclohexane,dimethylformamide,methylene chloride,pyridine,tetrahydrofuran,toluene,xylene. Insoluble in Acetone,butane,ethanol,ether,methanol,hexane,phenol,propanol,water.
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES *-CC(-*)C1=CC=CC=C1
IUPAC Name styrene
Molecular Weight (g/mol) 104.15
PubChem CID 7501
ChEBI CHEBI:27452
Concentration or Composition (by Analyte or Components) 10 wt% dispersion in water
Chemical Name or Material Polystyrene latex microsphere
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.