Learn More
Corning™ Polystyrene Microplates
Designed for in vitro diagnostic assays
Brand: Corning™ 2482
Code : 67
Additional Details : Weight : 0.04934kg
Description
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.- Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
- Recommended working volume of 50μL to 125μL
- Designed for in vitro diagnostic assays
- Untreated surface is hydrophobic in nature
- Color coded strips
- Bar coding available on request
Specifications
Strip Plate | |
Medium Binding | |
Polystyrene | |
75 to 125 μL | |
190 μL | |
25 |
8 x 12 | |
Black | |
96 | |
Non-treated | |
100Case | |
Black |