missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB Polysucrose for Cellular and Molecular Separations
Description
- Product Overview: Polysucrose 1000 kDa is a neutral, highly branched polysaccharide used for cellular and molecular separations. With high water solubility and biocompatibility, it is widely applied in gradient separation, size exclusion chromatography, and barrier diffusion studies.
- Features:
- High Molecular Weight: Approximately 1000 kDa for modeling large macromolecule behavior.
- Water-Soluble and Inert: Provides minimal interaction with biological components.
- Neutral Charge: Reduces nonspecific binding during separation or diffusion processes.
- Applications:
- Cell separation by density gradients.
- Molecular sieving and size exclusion studies.
- Modeling extracellular matrix-like diffusion behaviors.
- Standard for calibrating large molecule diffusion assays.
Specifications
Specifications
| Quantity | 1 g |
| Molecular Weight (g/mol) | 100 kDa |
| Grade | Research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?