missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ POLYTRIL™ Gloves

Product Code. 15806705
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15806705

Brand: Honeywell™ 223223007

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Honeywell™ POLYTRIL™ Gloves for handling of fine objects in a greasy, damp or soiled atmosphere.

  • White polyamide knitted glove
  • Gray oilproof nitrile coating on palm and fingertips
  • Elastic knitted wrist
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back improves the glove's breathability
  • The knitted and elasticated wrist gives the glove good support
  • The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
  • Its gray coating makes the glove more soilproof, thus increasing its last
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards
Description 13 gauge
Coating Material Polyamide, Nitrile
Color Gray, White
Size 7
Thickness (Metric) 0.25 mm
Length (Metric) 24.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.