Learn More
LABPLAS Polyurethane Sani-Sponge with Dey-Engley Neutralizing Broth
Product Code. 17976679
Change view
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17976679 None Pack of 100
17956509 Gloves Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17976679 Supplier LABPLAS Supplier No. KSS67810DE
  • ONLINE EXCLUSIVE PRICE
    250.56 €
    221.79 € / Pack of 100
    Save 28.77 €
    11% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
  • Polyurethane Sani-Sponge
  • Saturated with Dey-Engley neutralizing broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes None