missing translation for 'onlineSavingsMsg'
Learn More

LABPLAS Polyurethane Sani-Stick with Letheen Broth

Product Code. 17916689
Change view
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17916689 Gloves Pack of 100
17906509 None Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17916689 Supplier LABPLAS Supplier No. KSS67915LT
  • ONLINE EXCLUSIVE PRICE
    347.00 €
    281.74 € / Pack of 100
    Save 65.26 €
    18% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.

  • Polyurethane Sani-Stick
  • Saturated with Letheen broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit

Specifications

Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes Gloves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.