Learn More
LABPLAS Polyurethane Sani-Stick with Neutralizing Buffer
Product Code. 17916509
Change view
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17916509 None Pack of 100
17946689 Gloves Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17916509 Supplier LABPLAS Supplier No. KSS67910NE
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
  • Polyurethane Sani-Stick
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Especificaciones
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes None
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.