missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyurethene Safety Bumper

Product Code. 11721975 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11721975 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11721975 Supplier Brady™ Supplier No. 225020
  • ONLINE EXCLUSIVE PRICE
    24.00 €
    23.08 € / Pack of 1
    Save 0.92 €
    3% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose the Polyurethene Safety Bumper to suit different applications - use to warn of potential danger or help protect impact injury.

  • Choose these safety bumpers to suit different applications.

Specifications

Description Safety Bumpers - 2-dimensional corner piece
For Use With (Application) Safety Identification
Length (English) 3.3 ft.
Length (Metric) 1 m
Packaging Quantity 1 Ea.
Color Black
Material Polyurethane
Type Safety Bumpers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.