missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight

Product Code. 11349617
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349617 25 g 25g
11339617 100 g 100g
11359617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349617 Supplier Thermo Scientific Alfa Aesar Supplier No. 041243.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Physical Form Solid
Solubility Information Soluble in water.
Quantity 25 g
Percent Purity 98 to 99%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, High molecular weight, Hydrolyzed
What is the average molecular weight of Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight (Cat. No. 041243.xx)?

Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight has a typical average molecular weight of 126,000 g/mol.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.