missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone-iodine complex

Product Code. 15412447
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412447 50 g 50g
15422447 250 g 250g
15432447 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412447 Supplier Thermo Scientific Alfa Aesar Supplier No. 045782.18
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol

Polyvinylpyrrolidone-iodine complex is used as a slow iodine release formulation that iodinates lipids and oxidizes cell compounds such as tyrosine side chains. It is utilized to study microbial decontamination of human donor eyes as well as inactivation of infectious hematopoietic necrosis virus. It is also used as a carbon nanotube coating agent to enhance water solubility. It serves as a topical antiseptic in surgery, as an aerosol for skin and mucous membrane infections and is used to prevent infection in wounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25655-41-8
Molecular Formula (C6H9NO)nI2
Molecular Weight (g/mol) 364.95
MDL Number MFCD00084483
InChI Key CPKVUHPKYQGHMW-UHFFFAOYSA-N
Synonym povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
PubChem CID 410087
SMILES II.*-CC(-*)N1CCCC1=O

Specifications

Color Brown-Yellow
Physical Form Powder
Melting Point 300°C (decomposition)
Merck Index 14,7698
Solubility Information Soluble in water,ethyl alcohol,isopropyl alcohol,polyethylene glycol and glycerol. Insoluble in chloroform,carbon tetrachloride,hexane,acetone and petroleum ether.
Quantity 50 g
IUPAC Name 1-ethenylpyrrolidin-2-one;molecular iodine
Chemical Name or Material Polyvinylpyrrolidone-iodine complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.