missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone K 30, MP Biomedicals™

Product Code. 11485372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485372 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485372 Supplier MP Biomedicals Supplier No. 0219545190
  • ONLINE EXCLUSIVE PRICE
    216.00 €
    204.75 € / 500g
    Save 11.25 €
    5% Off
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 9003-39-8
Color White
Density 1.23-1.29g/mL
Molecular Formula (C6H9NO)n
MDL Number MFCD01076626
Quantity 500 g
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILES *-CC(-*)N1CCCC1=O
IUPAC Name 1-ethenylpyrrolidin-2-one
Molecular Weight (g/mol) 111.14
PubChem CID 6917
ChEBI CHEBI:82551
Formula Weight 40000
Physical Form Powder
Chemical Name or Material Polyvinylpyrrolidone K 30
Show More Show Less
compliance-icons
Product Identifier
  • Polyvinylpyrrolidone (PVP 40)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polyvinylpyrrolidone(PVP 40)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.