missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 360,000

Product Code. 15433989
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433989 1 kg 1kg
15413989 250 g 250g
15423989 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433989 Supplier Thermo Scientific Alfa Aesar Supplier No. J61381.A1
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
Physical Form Powder
Merck Index 14,7697
Solubility Information Soluble in water.
Quantity 1 kg
Formula Weight 360 kd
Chemical Name or Material Polyvinylpyrrolidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.