missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 8,000

Product Code. 11308768
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11308768 100 g 100g
11318768 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11308768

Brand: Thermo Scientific Alfa Aesar 041626.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as a cryoprotectant. It is used as a binder for tablets in pharmaceuticals. It is added to iodine and forms a complex povidone-iodine, which is having disinfectant properties, used in various products like solutions, ointment and pessaries. It is used as an adhesive in glue stick, emulsifier, additive for batteries, ceramics, fiberglass, inks, and inkjet paper, thickening agent in tooth whitening gels and to increase resolution in photoresists for cathode ray tubes. Further, it is used in personal care products such as shampoos, toothpastes, hair sprays and hair gels. In addition to this, it finds application in contact lens solutions, disintegrant for solution polymerization and in steel-quenching solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Physical Form Powder
Melting Point 130°C
Merck Index 14,7697
Solubility Information Soluble in water,alcohol,chlorinated hydrocarbons,amines,nitro paraffins,lower wt fatty acids and chloroform. Insoluble in ether.
Quantity 100 g
Odor Odorless
Chemical Name or Material Polyvinylpyrrolidone, M.W. 8,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.