missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, MP Biomedicals

Product Code. 11475372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475372 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475372 Supplier MP Biomedicals Supplier No. 0219545180
  • ONLINE EXCLUSIVE PRICE
    63.15 €
    59.85 € / 100g
    Save 3.30 €
    5% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity USP grade polyvinylpyrrolidone powder for laboratory and research applications.

Specifications

CAS 9003-39-8
Quantity 100 g
Molecular Formula (C6H9NO)n
MDL Number MFCD01076626
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILES *-CC(-*)N1CCCC1=O
IUPAC Name 1-ethenylpyrrolidin-2-one
Molecular Weight (g/mol) 111.14
PubChem CID 6917
ChEBI CHEBI:82551
Formula Weight 40,000 (Average )
Chemical Name or Material Polyvinylpyrrolidone
Show More Show Less
compliance-icons
Product Identifier
  • Polyvinylpyrrolidone (PVP 40)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polyvinylpyrrolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.