Learn More
Bohlender™ BOLA™ POM Globe Stirrer Couplings
Use these couplings with centrifuges at speeds of up to 1,200 rounds per minute. Bohlender™ BOLA™ POM Globe Stirrer Couplings feature good mechanical strength and powerful transmission of up to 300 Ncm.
Brand: Bohlender™ C398-12
Code : A0
Additional Details : Weight : 0.09900kg
Description
- Very low centrifugal forces due to low weight
- Suitable for both left- and right-handed rotation (except for GL 10 thread: no left-handed rotation possible)
- No resonance
- Simple assembly by means of screw joints with clamp rings
- Pivot (length 90mm) can be shortened by the user
- Maximum misalignment of axes 10mm
Specifications
8 mm | |
190 mm | |
Coupling Stirrer |
Ideal For Balancing Misalignment of Axis Between Agitator and Stirrer Shaft | |
1 Pack |