missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ POM Globe Stirrer Couplings
Use these couplings with centrifuges at speeds of up to 1,200 rounds per minute. Bohlender™ BOLA™ POM Globe Stirrer Couplings feature good mechanical strength and powerful transmission of up to 300 Ncm.
Brand: Bohlender™ C398-08
121.35 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Very low centrifugal forces due to low weight
- Suitable for both left- and right-handed rotation (except for GL 10 thread: no left-handed rotation possible)
- No resonance
- Simple assembly by means of screw joints with clamp rings
- Pivot (length 90mm) can be shortened by the user
- Maximum misalignment of axes 10mm
Specifications
6.5 mm | |
190 mm | |
Stirrer Coupling |
Ideal For Balancing Misalignment of Axis Between Agitator and Stirrer Shaft | |
1 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction