missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Pomalidomide

Product Code. 18796258
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796258 Supplier R&D Systems™ Supplier No. 6302/10
  • ONLINE EXCLUSIVE PRICE
    192.35 €
    166.95 € / 10mg
    Save 25.40 €
    13% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cereblon binder; also TNF-α inhibitor and antiangiogenic

Pomalidomide is a potent TNF-α inhibitor (IC50 = 13 nM). Also potently inhibits IL-2 (EC50 = 8 nM). Thalidomide derivative. Binds cereblon and inhibits its ubiquitination. Exhibits antiproliferative effects in a Namalwa lymphoma cell line. Antiangiogenic. Promotes degradation of transcription factor SALL4.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
CAS 19171-19-8
Assay Percent Range >98%
Quantity 10 mg
Product Line Tocris
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C13H11N3O4
Formula Weight Observed MW: 273.24
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.