missing translation for 'onlineSavingsMsg'
Learn More

Pomalidomide, MedChemExpress

Product Code. 16484328
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484328 10 mg 10mg
16465246 100 mg 100mg
16457047 5 g 5g
16440657 50 mg 50mg
16421139 1 g 1g
16411596 500 mg 500mg
16411016 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16484328 Supplier Medchem Express Supplier No. HY10984
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.

Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.

Specifications

Chemical Name or Material Pomalidomide
CAS 19171-19-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H11N3O4
Quantity 10 mg
Synonym CC-4047
Solubility Information DMSO : 50 mg/mL (182.99 mM; Need ultrasonic)
SMILES O=C1N(C(C2=C1C=CC=C2N)=O)C(C(N3)=O)CCC3=O
Molecular Weight (g/mol) 273.24
Formula Weight 273.24
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.