Learn More
Tocris Bioscience™ Pomalidomide
GREENER_CHOICE
Cereblon binder; also TNF-α inhibitor and antiangiogenic
Brand:  Tocris Bioscience™ 6302/10
Encompass
Product Code. 15746399
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Pomalidomide is a potent TNF-α inhibitor (IC50 = 13 nM). Also potently inhibits IL-2 (EC50 = 8 nM). Thalidomide derivative. Binds cereblon and inhibits its ubiquitination. Exhibits antiproliferative effects in a Namalwa lymphoma cell line. Antiangiogenic. Promotes degradation of transcription factor SALL4.
Specifications
Specifications
4-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
19171-19-8
>98%