missing translation for 'onlineSavingsMsg'
Learn More

Pomalidomide, TRC

Product Code. 30134838
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134838 250 mg 250mg
30133173 1 g 1g
18062020 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134838 Supplier Toronto Research Chemicals Supplier No. TRCP688200250MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13 H11 N3 O4
Molecular Weight (g/mol) 273.24
Synonym 1H-Isoindole-1,3(2H)-dione, 4-amino-2-(2,6-dioxo-3-piperidinyl)-, Phthalimide, 3-amino-N-(2,6-dioxo-3-piperidyl)- (8CI), 4-Amino-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione, 2-(2,6-Dioxopiperidin-3-yl)-4-amino-isoindole-1,3-dione, 4-Amino-2-(2,6-dioxo-3-piperidinyl)isoindole-1,3-dione, 4-Amino-2-(2,6-dioxo-3-piperidyl)isoindoline-1,3-dione, 4-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione, 4-Amino-2-(2,6-dioxopiperidine-3-yl)isoindoline-1,3-dione, Actimid, CC 4047, Pomalidomide, Pomalyst
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)isoindole-1,3-dione
SMILES Nc1cccc2C(=O)N(C3CCC(=O)NC3=O)C(=O)c12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11N3O4/c14-7-3-1-2-6-10(7)13(20)16(12(6)19)8-4-5-9(17)15-11(8)18/h1-3,8H,4-5,14H2,(H,15,17,18)
Formula Weight 273.075
Chemical Name or Material Pomalidomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.